Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ace-Thred
™
Thermometer/Tubing Adapter
Products
Product Category
adapters (15)
Brand
Sigma-Aldrich (1)
Manufacturer
ID (mm)
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
Z125350
Ace-Thred
™
Thermometer/Tubing Adapter,
Straight, joint: ST/NS 10/18, thread I.D. 6.5 mm
Expand
View Pricing
Z214337
Ace-Thred
™
Thermometer/Tubing Adapter,
With 2mm bore PTFE stopcock plug and hose connection, joint: ST/NS 24/40, thread I.D. 7.0 mm
Expand
View Pricing
Z219320
Ace-Thred
™
Thermometer/Tubing Adapter,
Twin angled, joint: ST/NS 24/25, thread I.D. 7.0 mm
Expand
View Pricing
Z125490
Ace-Thred
™
Thermometer/Tubing Adapter,
Angled and offset, joint: ST/NS 24/40, thread I.D. 7.0 mm
Expand
View Pricing
Z125369
Ace-Thred
™
Thermometer/Tubing Adapter,
Straight, joint: ST/NS 14/20, thread I.D. 7.0 mm
Expand
View Pricing
Z125385
Ace-Thred
™
Thermometer/Tubing Adapter,
Straight, joint: ST/NS 24/40, thread I.D. 7.0 mm
Expand
View Pricing
Z214396
Ace-Thred
™
Thermometer/Tubing Adapter,
Twin angled, side joint: ST/NS 24/40, top joint: ST/NS 24/40, bottom joint: ST/NS 24/25, thread I.D. 7.0 mm
Expand
View Pricing
Z125539
Ace-Thred
™
Thermometer/Tubing Adapter,
With hose connection, joint: ST/NS 24/40, thread I.D. 7.0 mm
Expand
View Pricing
Z125377
Ace-Thred
™
Thermometer/Tubing Adapter,
Straight, joint: ST/NS 19/22, thread I.D. 7.0 mm
Expand
View Pricing
Z125520
Ace-Thred
™
Thermometer/Tubing Adapter,
With hose connection, joint: ST/NS 19/22, thread I.D. 7.0 mm
Expand
View Pricing
Z125512
Ace-Thred
™
Thermometer/Tubing Adapter,
With hose connection, joint: ST/NS 14/20, thread I.D. 7.0 mm
Expand
View Pricing
Z214361
Ace-Thred
™
Thermometer/Tubing Adapter,
With hose connection, joint: ST/NS 24/40, thread I.D. 9.0-10.5 mm
Expand
View Pricing
Z125458
Ace-Thred
™
Thermometer/Tubing Adapter,
Straight, joint: ST/NS 24/40, thread I.D. 9.0-10.5 mm
Expand
View Pricing
Z214388
Ace-Thred
™
Thermometer/Tubing Adapter,
With hose connection, joint: ST/NS 29/42, thread I.D. 9.0-10.5 mm
Expand
View Pricing
Z125547
Ace-Thred
™
Thermometer/Tubing Adapter,
For electrode with hose connection, joint: ST/NS 24/40, thread I.D. 12.5-14.0 mm
Expand
View Pricing