
Acequinocyl

Synonym(s):
2-(Acetyloxy)-3-dodecyl-1,4-naphthalenedione, 3-Acetoxy-2-dodecyl-1,4-naphthoquinone, 3-Dodecyl-1,4-dihydro-1,4-dioxo-2-naphthyl acetate
Empirical Formula (Hill Notation):
C24H32O4
CAS Number:
Molecular Weight:
384.51
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2