Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
8-Hydroxy-7-iodo-5-quinolinesulfonic acid
Synonym(s):
Ferron, Iodoxyquinolinesulfonic acid
Empirical Formula (Hill Notation):
C
9
H
6
INO
4
S
CAS Number:
547-91-1
Molecular Weight:
351.12
EC Number:
208-938-4
Products
Brand
Supelco (1)
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Special Grade
Puriss (1)
Puriss P.A. (1)
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
55370
8-Hydroxy-7-iodo-5-quinolinesulfonic acid,
for spectrophotometric det. of Fe(III), ≥98.5%
Expand
View Pricing