
8-Geranyloxypsoralen

Synonym(s):
9-[[(2E)-3,7-Dimethyl-2,6-octadien-1-yl]oxy]-7H-furo[3,2-g][1]benzopyran-7-one
Empirical Formula (Hill Notation):
C21H22O4
CAS Number:
Molecular Weight:
338.40
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing