
6-Phosphogluconic Dehydrogenase from yeast

Synonym(s):
6-Phospho-D-gluconate:NADP+ 2-oxidoreductase (decarboxylating), 6-Phosphogluconic Dehydrogenase from yeast
CAS Number:
Products
Brand
Biological Source
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing