6-Bromo-3-pyridinylboronic acid
Synonym(s):
2-Bromopyridine-5-boronic acid
Linear Formula:
Br(C5H3N)B(OH)2
CAS Number:
Molecular Weight:
201.81
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing