Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
6-Benzylaminopurine
Synonym(s):
6-BAP, BA, N
6
-Benzyladenine, 6-Benzylaminopurine
Empirical Formula (Hill Notation):
C
12
H
11
N
5
CAS Number:
1214-39-7
Molecular Weight:
225.25
EC Number:
214-927-5
Products
Product Category
bioactive small molecules (1)
building blocks (1)
Brand
Sigma-Aldrich (3)
Supelco (1)
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
B3408
6-Benzylaminopurine,
suitable for plant cell culture
Expand
View Pricing
13151
6-Benzylaminopurine,
ReagentPlus
®
, ≥99.0% (HPLC)
Expand
View Pricing
200241
6-Benzylaminopurine,
Plant cell division factor.
Expand
View Pricing
67619
6-Benzylaminopurine,
PESTANAL
®
, analytical standard
Expand
View Pricing