Skip to Content
Merck
5,6-Dihydro-2H-pyran-2-one technical grade, 90%

5,6-Dihydro-2H-pyran-2-one

5,6-Dihydro-2H-pyran-2-one technical grade, 90%
Empirical Formula (Hill Notation):
C5H6O2
CAS Number:
Molecular Weight:
98.10

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing