
5-(2-Hydroxyethyl)-4-methylthiazole

Synonym(s):
4-Methyl-5-thiazoleethanol, 5-(2-Hydroxyethyl)-4-methylthiazole, Sulfurol
Empirical Formula (Hill Notation):
C6H9NOS
CAS Number:
Molecular Weight:
143.21
Products
Brand
Boiling Point (°C)
Formula Weight
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing