
4-Maleimidobutyric acid

Synonym(s):
γ-Maleimidobutyric acid, 4-Maleimidobutanoic acid, N-(3-Carboxypropyl)maleimide, N-Maleoyl-4-aminobutyric acid, N-Maleoyl-GABA
Empirical Formula (Hill Notation):
C8H9NO4
CAS Number:
Molecular Weight:
183.16
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing