Skip to Content
Merck
4-(Diphenylhydroxymethyl)benzoic acid ≥98.0% (HPLC)

4-(Diphenylhydroxymethyl)benzoic acid

4-(Diphenylhydroxymethyl)benzoic acid ≥98.0% (HPLC)
Synonym(s):
4-(α-Hydroxybenzhydryl)benzoic acid, α,α-Diphenyl-α-hydroxy-p-toluic acid
Empirical Formula (Hill Notation):
C20H16O3
CAS Number:
Molecular Weight:
304.34

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing