![4′-Bromo-2,2,2-trifluoroacetophenone ≥98%](/deepweb/assets/sigmaaldrich/product/structures/286/540/6e4a8199-c14d-4e16-af1b-73508e1ebfe4/640/6e4a8199-c14d-4e16-af1b-73508e1ebfe4.png)
4′-Bromo-2,2,2-trifluoroacetophenone
![4′-Bromo-2,2,2-trifluoroacetophenone ≥98%](/deepweb/assets/sigmaaldrich/product/structures/286/540/6e4a8199-c14d-4e16-af1b-73508e1ebfe4/640/6e4a8199-c14d-4e16-af1b-73508e1ebfe4.png)
Synonym(s):
4-Bromo-α,α,α-trifluoroacetophenone
Linear Formula:
BrC6H4COCF3
CAS Number:
Molecular Weight:
253.02
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing