4-Amino-3-hydroxybenzoic acid
Linear Formula:
H2NC6H3(OH)CO2H
CAS Number:
Molecular Weight:
153.14
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing