
4-Amino-1-Boc-piperidine

Synonym(s):
1-Boc-4-piperidinamine, tert-Butyl 4-amino-1-piperidinecarboxylate
Empirical Formula (Hill Notation):
C10H20N2O2
CAS Number:
Molecular Weight:
200.28
Products
Product Category
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing