Skip to Content
Merck
3-Chloro-4-fluoroaniline 98%

3-Chloro-4-fluoroaniline

3-Chloro-4-fluoroaniline 98%
Synonym(s):
3-Chloro-4-fluoroaniline
Linear Formula:
3-(Cl),4-(F)C6H3NH2
CAS Number:
Molecular Weight:
145.56

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing