
3,3′,5′-Triiodo-L-thyronine (Reverse T3) solution

Synonym(s):
3,3′,5′-Triiodo-L-thyronine, Reverse T3, rT(3)
Empirical Formula (Hill Notation):
C15H12I3NO4
CAS Number:
Molecular Weight:
650.97
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing