Skip to Content
Merck
3,3′,5,5′-Tetramethylbenzidine dihydrochloride tablet, 1 mg substrate per tablet

3,3′,5,5′-Tetramethylbenzidine dihydrochloride

3,3′,5,5′-Tetramethylbenzidine dihydrochloride tablet, 1 mg substrate per tablet
Synonym(s):
4,4′-Diamino-3,3′,5,5′-tetramethylbiphenyl dihydrochloride
Empirical Formula (Hill Notation):
C16H20N2 · 2HCl
CAS Number:
Molecular Weight:
313.27
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing