Skip to Content
Merck
2-tert-Butylimino-2-diethylamino-1,3-dimethylperhydro-1,3,2-diazaphosphorine purum, ≥98.0% (GC)

2-tert-Butylimino-2-diethylamino-1,3-dimethylperhydro-1,3,2-diazaphosphorine

2-tert-Butylimino-2-diethylamino-1,3-dimethylperhydro-1,3,2-diazaphosphorine purum, ≥98.0% (GC)
Synonym(s):
BEMP
Empirical Formula (Hill Notation):
C13H31N4P
CAS Number:
Molecular Weight:
274.39

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1