
2-(Methylthio)ethylamine

Synonym(s):
2-Aminoethyl methyl sulfide, S-Methylcysteamine
Linear Formula:
CH3SCH2CH2NH2
Products
Product Category
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing