
2-Formylphenylboronic acid

Synonym(s):
2-(Dihydroxyboryl)benzaldehyde, 2-Boronobenzaldehyde, 2-Formylbenzeneboronic acid, o-Formylbenzeneboronic acid, o-Formylphenylboronic acid, NSC 157839
Linear Formula:
HCOC6H4B(OH)2
CAS Number:
Molecular Weight:
149.94
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing