Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2-Ethylhexyl acrylate
Synonym(s):
(±)-Acrylic acid 2-ethylhexyl ester, 2-Ethylhexyl acrylate
Linear Formula:
CH
2
=CHCOOCH
2
CH(C
2
H
5
)(CH
2
)
3
CH
3
CAS Number:
103-11-7
Molecular Weight:
184.28
EC Number:
203-080-7
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (2)
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Monomer Type
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
290815
2-Ethylhexyl acrylate,
98%, contains ≥0.001-≤0.11% monomethyl ether hydroquinone as stabilizer
Expand
View Pricing
8.00835
2-Ethylhexyl acrylate,
(stabilised with hydroquinone monomethyl ether) for synthesis
Expand
View Pricing
67262
2-Ethylhexyl acrylate,
analytical standard
Expand
View Pricing