Skip to Content
Merck
2-(2-Chloroethyl)-1-methylpyrrolidine hydrochloride 99%

2-(2-Chloroethyl)-1-methylpyrrolidine hydrochloride

2-(2-Chloroethyl)-1-methylpyrrolidine hydrochloride 99%
Empirical Formula (Hill Notation):
C7H14ClN · HCl
CAS Number:
Molecular Weight:
184.11
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing