
1-(Trimethylsilyl)imidazole - Pyridine mixture

Synonym(s):
Silylating mixture VII, TMSI+Pyridine
CAS Number:
Products
Brand
Physical Form
Quality Segment
Reaction Type
Reagent Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing