
1-Methylnicotinamide chloride

Synonym(s):
3-Carbamyl-1-methylpyridinium chloride, MNA, N-Methylnicotinic acid amide chloride, N1-Methylnicotinamide chloride
Linear Formula:
C7H9N2OCl
CAS Number:
Molecular Weight:
172.61
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing