Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1-Fluoro-2,4-dinitrobenzene
Synonym(s):
2,4-Dinitro-1-fluorobenzene, DNFB, DNPF, FDNB, Sanger reagent
Linear Formula:
(O
2
N)
2
C
6
H
3
F
CAS Number:
70-34-8
Molecular Weight:
186.10
EC Number:
200-734-3
Products
Product Category
building blocks (1)
substrates (1)
Brand
Sigma-Aldrich (2)
Supelco (1)
Boiling Point (°C)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Purum (1)
Purum P.A. (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
D1529
1-Fluoro-2,4-dinitrobenzene,
≥99%
Expand
View Pricing
42085
1-Fluoro-2,4-dinitrobenzene,
purum p.a., ≥98.0% (GC)
Expand
View Pricing
73177
1-Fluoro-2,4-dinitrobenzene,
for HPLC derivatization, LiChropur
™
, ≥99.0% (GC)
Expand
View Pricing