Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1-(Chloromethyl)naphthalene
Synonym(s):
1-Naphthylmethyl chloride
Linear Formula:
C
10
H
7
CH
2
Cl
CAS Number:
86-52-2
Molecular Weight:
176.64
EC Number:
201-678-2
Products
Product Category
building blocks (2)
Brand
Sigma-Aldrich (2)
Boiling Point (°C)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Technical (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
C54201
1-(Chloromethyl)naphthalene,
90%
Expand
View Pricing
25170
1-(Chloromethyl)naphthalene,
technical, ≥97.0% (GC)
Expand
View Pricing