
1,3-Propanediol

Synonym(s):
1,3-Dihydroxypropane, Trimethylene glycol, 1,3-Propanediol
Linear Formula:
HO(CH2)3OH
CAS Number:
Molecular Weight:
76.09
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade