Skip to Content
Merck
1,2-Dipalmitoyl-rac-glycerol ≥99%

1,2-Dipalmitoyl-rac-glycerol

1,2-Dipalmitoyl-rac-glycerol ≥99%
Synonym(s):
(±)-1,2-Dipalmitin, 1,2-Dihexadecanoyl-rac-glycerol, rac-Glycerol 1,2-dipalmitate, DL-1,2-Dipalmitin, DG(16:0/16:0/0:0)[rac], Hexadecanoic acid 1,1′-[1-(hydroxymethyl)-1,2-ethanediyl] ester
Empirical Formula (Hill Notation):
C35H68O5
CAS Number:
Molecular Weight:
568.91
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing