Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ribonucleic acid from torula yeast
Synonym(s):
RNA, Ribonucleic Acid, Yeast, Ribonucleic acid from torula yeast
CAS Number:
63231-63-0
Products
Product Category
nucleic acids (5)
Brand
Millipore (1)
Sigma-Aldrich (4)
Color
Physical Form
Quality Segment
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
R6625
Ribonucleic acid from torula yeast,
Type VI
Expand
View Pricing
R6750
Ribonucleic acid from torula yeast,
Expand
View Pricing
55714
Ribonucleic acid from torula yeast,
Expand
View Pricing
R3629
Ribonucleic acid from torula yeast,
Type IX
Expand
View Pricing
R6875
Ribonucleic acid from torula yeast,
core, Type II-C
Expand
View Pricing