![1,10-Phenanthroline monohydrate GR ACS](/deepweb/assets/sigmaaldrich/product/images/306/245/6ddea61b-a9bc-4b3c-97f8-0c8b89df9136/640/6ddea61b-a9bc-4b3c-97f8-0c8b89df9136.jpg)
1,10-Phenanthroline monohydrate
![1,10-Phenanthroline monohydrate GR ACS](/deepweb/assets/sigmaaldrich/product/images/306/245/6ddea61b-a9bc-4b3c-97f8-0c8b89df9136/640/6ddea61b-a9bc-4b3c-97f8-0c8b89df9136.jpg)
Synonym(s):
1,10-Phenanthroline monohydrate
Empirical Formula (Hill Notation):
C12H8N2 · xH2O
CAS Number:
Molecular Weight:
180.21 (anhydrous basis)
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing