Skip to Content
Merck
1,10-Diiododecane 95%

1,10-Diiododecane

1,10-Diiododecane 95%
Synonym(s):
Decamethylene diiodide
Linear Formula:
I(CH2)10I
CAS Number:
Molecular Weight:
394.07
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing