Skip to Content
Merck
All Photos(1)

Key Documents

1353500

USP

Isotretinoin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

13-cis-Retinoic acid, 13-cis-Vitamin A acid, Isotretinoin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O2
CAS Number:
Molecular Weight:
300.44
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

isotretinoin

manufacturer/tradename

USP

mp

172-175 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H]\C(C(O)=O)=C(C)\C=C\C=C(C)\C=C\C1=C(C)CCCC1(C)C

InChI

1S/C20H28O2/c1-15(8-6-9-16(2)14-19(21)22)11-12-18-17(3)10-7-13-20(18,4)5/h6,8-9,11-12,14H,7,10,13H2,1-5H3,(H,21,22)/b9-6+,12-11+,15-8+,16-14-

InChI key

SHGAZHPCJJPHSC-XFYACQKRSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Isotretinoin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Isotretinoin Capsules
  • Tretinoin

Biochem/physiol Actions

13-cis-Retinoic acid (RA) has anti-inflammatory and anti-tumor action. The action of RA is mediated through RAR-β and RAR-α receptors. RA attenuates iNOS expression and activity in cytokine-stimulated murine mesangial cells. It induces mitochondrial membrane permeability transition, observed as swelling and as a decrease in membrane potential, and stimulates the release of cytochrome c implicating mechanisms through the apoptosis pathway. These activities are reversed by EGTA and cyclosporin A. RA also increases MMP-1 protein expression partially via increased transcription.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isotretinoin
United States Pharmacopeia, 2457-2457 (2020)
Tretinoin
United States Pharmacopeia, 41(4), 4478-4478 (2020)
Isotretinoin Capsules
United States Pharmacopeia, 48(4), 2458-2458 (2023)
The role and mechanism of action of 13-cis-retinoic acid in the treatment of severe (nodulocystic) acne.
D H Jones
Pharmacology & therapeutics, 40(1), 91-106 (1989-01-01)
H J M J Crijns et al.
The British journal of dermatology, 164(2), 238-244 (2010-08-19)
Most of the publications on isotretinoin, pregnancy and compliance with the pregnancy prevention programme (PPP) originate from North America. Information specific for the European situation is very limited. The aim of this study was to identify publications describing the use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service