Skip to Content
Merck
All Photos(1)

Key Documents

1329913

USP

5-Hydroxy-L-tryptophan

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

L-2-Amino-3-(5-hydroxyindolyl)propionic acid, L-5-HTP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2O3
CAS Number:
Molecular Weight:
220.22
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

tryptophan

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

N[C@@H](Cc1c[nH]c2ccc(O)cc12)C(O)=O

InChI

1S/C11H12N2O3/c12-9(11(15)16)3-6-5-13-10-2-1-7(14)4-8(6)10/h1-2,4-5,9,13-14H,3,12H2,(H,15,16)/t9-/m0/s1

InChI key

LDCYZAJDBXYCGN-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

5-Hydroxy-L-tryptophan USP reference standard suitable for use in specified USP compendial quality tests and assays.

Also used to prepare standard solution for the impurity analysis of 5-Hydroxy-L-tryptophan by using liquid chromatography coupled with UV detector.

Biochem/physiol Actions

Immediate precursor of serotonin; L-aromatic amino acid decarboxylase substrate.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

5-Hydroxy-l-tryptophan
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(2), 5313-5313 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service