Skip to Content
Merck
All Photos(1)

Documents

Y0002163

Testosterone for ID and assay

CRS, European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Testosterone, 17β-Hydroxy-3-oxo-4-androstene, 17β-Hydroxy-4-androsten-3-one, 4-Androsten-17β-ol-3-one, trans-Testosterone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H28O2
CAS Number:
Molecular Weight:
288.42
Beilstein:
1915399
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

Quality Level

API family

testosterone

manufacturer/tradename

EDQM

storage temp.

2-8°C

SMILES string

C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2O

InChI

1S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h11,14-17,21H,3-10H2,1-2H3/t14-,15-,16-,17-,18-,19-/m0/s1

InChI key

MUMGGOZAMZWBJJ-DYKIIFRCSA-N

Gene Information

human ... AR(367)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Testosterone for ID and assay EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Testosterone secreted by the testis is converted to dihydrotestosterone in the target tissues where it appears to mediate many of the biological actions of testosterone. Androgens direct the development of the male phenotype during embryogenesis and at puberty.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service