Skip to Content
Merck
All Photos(1)

Documents

CRM34371

Supelco

Propetamphos

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20NO4PS
CAS Number:
Molecular Weight:
281.31
Beilstein:
1979853
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

application(s)

environmental

format

neat

storage temp.

−20°C

SMILES string

CCNP(=S)(OC)O\C(C)=C\C(=O)OC(C)C

InChI

1S/C10H20NO4PS/c1-6-11-16(17,13-5)15-9(4)7-10(12)14-8(2)3/h7-8H,6H2,1-5H3,(H,11,17)/b9-7+

InChI key

BZNDWPRGXNILMS-VQHVLOKHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.Download your certificate at: http://www.sigma-aldrich.com

Application

Propetamphos is an insecticide and acaricide used in public health and livestock situation against cockroaches, flies, ants, moths, fleas, mosquitoes and termites. Under Reg. (EC) No 1107/2009 It is not approved for use in the European Union (EU). A default maximum residue level (MRL) of 0.01 mg/kg has been set according to ART 18(1)(b) Reg 396/2005 . The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service