Skip to Content
Merck
All Photos(1)

Key Documents

BOTB9052

Supelco

Frangula purshiana Bark

Botanical reference material

Sign Into View Organizational & Contract Pricing


About This Item

NACRES:
NA.24

manufacturer/tradename

BOTANICERT®

application(s)

food and beverages
pharmaceutical

format

matrix material

storage temp.

2-25°C

General description

English name: Cascara buckthorn, French name: Cascara

This botanical reference material (BRM) is manufactured by BotaniCERT™ and EXTRASYNTHESE™. The material is sourced from botanical gardens, collected by botanical experts during the flowering period, analyzed by seasoned phytochemists and geneticians.

BotaniCERT Botanical Reference Materials are samples of plants or plant parts (leaf, root, rhizome, fruit, flower, seed...) authenticated by organoleptic criteria, macroscopic and microscopic examination, HP-TLC and HPLC phytochemical fingerprint and genetic markers by PCR. They are given with 2 or 3 markers (ITS, matK, rbcL) and DNA sequences are reported.

The products are provided as 5g samples and a detailed Certificate of Analysis, including related pictures, chromatograms and DNA sequences is available.

Using a Botanical Reference Material in an identification analysis allows calibration of methods and increases the guarantee of the result obtained.

Application

For identity and authenticity testing of botanical materials

Legal Information

BOTANICERT is a registered trademark of BOTANICERT, Société, FR

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service