Skip to Content
Merck
All Photos(1)

Documents

59660-U

Supelco

Supelguard HPLC Guard Cartridge Holder

Stand-alone, for use with Supelguard cartridges (2 cm L. x 2.1 to 4.0 mm I.D.)

Synonym(s):

HPLC Guard Cartridge System

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

product name

Supelguard Guard Cartridge Holder, Stand-alone, for use with Supelguard cartridges (2 cm L. x 2.1 to 4.0 mm I.D.)

packaging

pkg of 1 ea fittings included

technique(s)

HPLC: suitable

compatibility

for use with Supelguard cartridges (2 cm L. x 2.1 to 4.0 mm I.D.)

Looking for similar products? Visit Product Comparison Guide

Footnote

For 2.1, 3.0, 4.0 and 4.6 mm I.D. Supelco columns.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

-methylcarbamate 10 μg/mL; Diuron; Propham; Siduron; Methiocarb, analytical standard; Linuron 10 μg/mL; Swep 10 μg/mL; Chlorpropham 10 μg/mL; Barban; Neburon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service