Skip to Content
Merck
All Photos(1)

Key Documents

57170-U

Supelco

ENVI-18 DSK SPE Disk

C18, diam. 90 mm, pk of 12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

product name

ENVI-18 DSK SPE Disk, diam. 90 mm, pk of 12

Agency

suitable for EPA 1613

packaging

pk of 12

technique(s)

solid phase extraction (SPE): suitable

diam.

90 mm

matrix active group

C18 bonding, octadecyl

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Retention Mechanism: Reversed-phase
Sample Matrix Compatibility: Aqueous solutions (drinking water)

  • The SPE membrane equivalents of ENVI-18 and ENVI-8 packed bed SPE sorbents
  • Porous glass fiber membranes embedded with C18 or C8 modified silica particles
  • Provides faster flow rates and exhibits less clogging than PTFE discs for the extraction of organic contaminants from drinking water samples
  • Typical applications include polynuclear aromatic hydrocarbons (PAHs), polychlorinated biphenyls (PCBs), phthalates, semivolatile organics, paraquat and diquat, pesticides and herbicides

Legal Information

ENVI is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Reversed-phase interaction will retain most molecules with hydrophobic character; it is very useful for extracting analytes that are very diverse in structure within the same sample.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service