Skip to Content
Merck
All Photos(8)

Documents

ZRB1981

Sigma-Aldrich

Anti-SMAD4 Antibody, clone 1C14 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C14, recombinant monoclonal

description

1C14 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 60.44 kDa
observed mol wt ~59 kDa

species reactivity

human, mouse

species reactivity (predicted by homology)

bovine, monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminus

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1C14 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects SMAD4. It targets an epitope within 16 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal region of human SMAD4.

Application

Quality Control Testing

Evaluated by Western Blotting in NIH3T3 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected SMAD4 in NIH3T3 cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected SMAD4 in HepG2 and A431 cell lysates.

Western Blotting KO Analysis: A 1:1,000 dilution from a representative lot detected SMAD4 in lysate from wild-type HeLa cells but not in SMAD4 knockout HeLa lysate.

Affinity Binding Assay: A representative lot of this antibody bound SMAD4 with a KD of 1.0 x 10-12 in an affinity binding assay.

ELISA: A representative lot of this antibody detected SMAD4 in ELISA application.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-SMAD4, clone 1C14 ZooMAb®, Cat. No. ZRB1981, is a recombinant Rabbit monoclonal antibody that specifically targets SMAD4 and is tested for use in Affinity Binding Assay, ELISA, Western Blotting, and Western Blotting using Knockout lysates.

Target description

Mothers against decapentaplegic homolog 4 (UniProt: Q13485; also known as MAD homolog 4, Mothers against DPP homolog 4, Deletion target in pancreatic carcinoma 4, SMAD family member 4, SMAD 4, Smad4, hSMAD4) is encoded by the SMAD4 (also known as DPC4, MADH4) gene (Gene ID: 4089) in human. SMAD proteins are signal transducers for the members of the transforming growth factor-b (TGF-b) superfamily. SMAD4 is a transcription factor that is involved in TGF-β signaling. In the absence of a ligand it is monomeric and is localized in the cytoplasm. Upon ligand binding it forms a heterotrimer with SMAD2 or SMAD3 and another molecule of SMAD4 to form the active complex that translocates to the nucleus. SMAD4 can undergo monoubiquitination on lysine 519 by E3 ubiquitin-protein ligase TRIM33 and this monoubiquitination hampers its ability to form a stable complex with activated SMAD2/3 resulting in inhibition of TGF-b signaling. Deubiquitination by USP9X is reported to restore its competence to mediate TGF-b signaling. As the main effector of TGF-β signaling, SMAD4 has been found to be non-functional in more than half of adenocarcinomas of the pancreatic duct and in colorectal cancers. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Chen, HB., et al. (2005). J. Biol. Chem. 280(22); 21329-21936; Kawabata, M., et al. (1998). EMBO J. 17(14); 4056-4065).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service