Skip to Content
Merck
All Photos(1)

Key Documents

Z744269

Wilmad® Suprasil® Aqueous Flow Cell

storage condition room temperature, pack of 1 ea

Synonym(s):

EPR aqueous cell, aqueous cell

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15

packaging

pack of 1 ea

manufacturer/tradename

Wilmad WG-804-Q

General description

Continuous flow is a powerful tool to study free radical intermediates with short life-time at steady state. The length of the flow tube is 130 mm before reaching the mixing chamber, dead volume is less than 9.5 µL, which corresponds to 2 ms dead time at 5 ml/sec flow rate. Inlet and outlet ports could be connected to 1/8” or 1/4” Tygon tubing depending on the size of the ports. Rinse the cell thoroughly after each usage to prevent clogging. WG-804-Q fits most Bruker TE102 and JEOL TE011 cavities. The sample thickness is 0.25 mm in the flat section.

Application

EPR

Features and Benefits

Reusable after rinsing

Legal Information

Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Wilmad is a registered trademark of SP Industries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service