Skip to Content
Merck
All Photos(1)

Key Documents

Z744005

SAFC

Worthington Accessories

necktube core for LD35 and LD50 dewars

Synonym(s):

Laboratory Accessories, Worthington Products

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122806
NACRES:
NB.51

packaging

pack of 1 ea

manufacturer/tradename

Worthington D050-9C16

technique(s)

cryopreservation: suitable

Looking for similar products? Visit Product Comparison Guide

General description

All Worthington cryostorage refrigerators and dewars each comes with a standard lid, so it is not necessary to purchase an additional one. However, many customers buy spare caps/necktube cores in order to modify them so that different kinds of sample storage devices can be suspended into a storage cannister.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service