Y1003
2x YT medium
Liquid microbial growth medium
Synonym(s):
2x Yeast Extract Tryptone medium, 2xTY medium, 2xYT Medium
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106225
NACRES:
NA.85
Recommended Products
grade
for molecular biology
Quality Level
sterility
sterile; 0.2 μm filtered
form
liquid
technique(s)
microbiological culture: suitable
pH
6.5-7.5
application(s)
microbiology
storage temp.
room temp
suitability
nonselective for Escherichia coli
nonselective for coliforms
General description
2xYT ( 2x Yeast Extract Tryptone medium) is a standard microbial growth medium used for the cultivation of E. coli and M13 bacteriophage. This nutrient-rich microbial broth contains peptides, amino acids, and water-soluble vitamins in a low-salt formulation.
Application
2x YT medium has been used for non-selective cultivation of M13 bacteriophage, E. coli strains for cloning , DNA plasmid production, and production of recombinant proteins. It is also suitable for selective cultivation when appropriate antibiotics are added.
Features and Benefits
2xYT liquid provides:
- Filter-sterilized, ready-to-use format
- Convenient package size
- Standard formulation
Components
16 g/L Tryptone
10 g/L Yeast Extract
5.0 g/L NaCl
10 g/L Yeast Extract
5.0 g/L NaCl
related product
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Victor V Kuprianov et al.
Virus research, 284, 197984-197984 (2020-04-24)
Hepatitis C virus (HCV) can cause chronic infection and evade the immune response. The generation and maintenance of an effective T-cell response is important for immune-mediated control of HCV infection. The purpose of this study was to obtain recombinant mosaic
Dynamical model fitting to a synthetic positive feedback circuit in E. coli
Tica J. et al.
Journal of Separation Science (2020)
Nicholas Collins et al.
Cell, 178(5), 1088-1101 (2019-08-24)
Mammals evolved in the face of fluctuating food availability. How the immune system adapts to transient nutritional stress remains poorly understood. Here, we show that memory T cells collapsed in secondary lymphoid organs in the context of dietary restriction (DR) but
Hong-Yi Kuo et al.
Glycobiology, 26(2), 155-165 (2015-10-09)
Galectin-3 is a ubiquitous lectin exerting multiple cellular functions such as RNA splicing, protein trafficking and apoptosis. Its expression is positively correlated with the poor prognosis in lung cancer patients. Galectin-3 can promote cancer progression through its effects on cell
Zehua Sun et al.
Frontiers in immunology, 13, 869825-869825 (2022-04-26)
Phage display is a well-established technology for in vitro selection of monoclonal antibodies (mAb), and more than 12 antibodies isolated from phage displayed libraries of different formats have been approved for therapy. We have constructed a large size (10^11) human
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service