Skip to Content
Merck
All Photos(1)

Documents

T0397

Sigma-Aldrich

L-Thyroxine sodium salt pentahydrate

γ-irradiated, powder, BioXtra, suitable for cell culture

Synonym(s):

3-[4-(4-Hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]-L-alanine sodium salt, Sodium levothyroxine, T4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H2I2OC6H2I2CH2CH(NH2)COONa · 5H2O
CAS Number:
Molecular Weight:
888.93
Beilstein:
5721845
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

sterility

γ-irradiated

product line

BioXtra

Assay

≥98%

form

powder

potency

5-50 ng/mL

technique(s)

cell culture | mammalian: suitable

mp

207-210 (dec.) (lit.)

solubility

cell culture medium: 0.1 mg/mL
H2O: soluble

storage temp.

−20°C

SMILES string

O.O.O.O.O.[Na+].N[C@@H](Cc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1)C([O-])=O

InChI

1S/C15H11I4NO4.Na.5H2O/c16-8-4-7(5-9(17)13(8)21)24-14-10(18)1-6(2-11(14)19)3-12(20)15(22)23;;;;;;/h1-2,4-5,12,21H,3,20H2,(H,22,23);;5*1H2/q;+1;;;;;/p-1/t12-;;;;;;/m0....../s1

InChI key

JMHCCAYJTTWMCX-QWPJCUCISA-M

Gene Information

mouse ... Cnr2(12802)
rat ... Cnr1(25248)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

L-Thyroxine (T4) and triiodo-L-thyronine (T3) are iodine-containing hormones produced from thyroglobulin in the thyroid follicular cells. The stimulation of metabolic rate and regulation of growth and development by these hormones appear to be due to their effects on DNA transcription and thus, protein synthesis.

Physical form

powder-0 °C; stock-frozen in working aliquots, avoid repeated freeze/thaw, solution stable 2-8 °C, 30 days

Reconstitution

To prepare 20 μg/ml stock solution, add 50 ml sterile culture medium.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1

Target Organs

Thyroid,Cardio-vascular system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giuseppe Ianiri et al.
mBio, 6(2) (2015-04-02)
Fungal diseases represent a major burden to health care globally. As with other pathogenic microbes, there is a limited number of agents suitable for use in treating fungal diseases, and resistance to these agents can develop rapidly. Cryptococcus neoformans is
Lilian Pukk et al.
Molecular ecology resources, 15(5), 1145-1152 (2015-02-24)
Massively parallel sequencing a small proportion of the whole genome at high coverage enables answering a wide range of questions from molecular evolution and evolutionary biology to animal and plant breeding and forensics. In this study, we describe the development
Mahdokht Jouiaei et al.
Molecular biology and evolution, 32(6), 1598-1610 (2015-03-12)
Despite Cnidaria (sea anemones, corals, jellyfish, and hydroids) being the oldest venomous animal lineage, structure-function relationships, phyletic distributions, and the molecular evolutionary regimes of toxins encoded by these intriguing animals are poorly understood. Hence, we have comprehensively elucidated the phylogenetic
Laam Li et al.
BMC genomics, 16, 637-637 (2015-08-27)
Legionella pneumophila (Lp) is a water-borne opportunistic pathogen. In water, Lp can survive for an extended period of time until it encounters a permissive host. Therefore, identifying genes that are required for survival in water may help develop strategies to
Pelin Sahlén et al.
Genome biology, 16, 156-156 (2015-08-28)
Although the locations of promoters and enhancers have been identified in several cell types, we still have limited information on their connectivity. We developed HiCap, which combines a 4-cutter restriction enzyme Hi-C with sequence capture of promoter regions. Applying the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service