Skip to Content
Merck
All Photos(2)

Key Documents

SRP0192

Sigma-Aldrich

PARP1 Active human

recombinant, expressed in baculovirus infected insect cells, ≥80% (SDS-PAGE)

Synonym(s):

ADPRT, NAD(+) ADP-ribosyltransferase 1, Poly (ADP-ribose) Polymerase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected insect cells

Assay

≥80% (SDS-PAGE)

form

aqueous solution

mol wt

140 kDa

packaging

pkg of 10 and 20 μg

manufacturer/tradename

Sigma-Aldrich

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.02 mg/mL

technique(s)

inhibition assay: suitable

NCBI accession no.

UniProt accession no.

application(s)

life science and biopharma

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PARP1(142)

General description

Research area: Cell Signaling

Human PARP1 (GenBank Accession No. NM_001618), full length with N-terminal GST tag, MW = 140 kDa, expressed in a Baculovirus infected Sf9 cell expression system. Poly [ADP-ribose] polymerase 1 (PARP1) belongs to the DNA-dependent nuclear enzyme superfamily. Its structure includes an N-terminal with three zinc finger DNA-binding domains, an auto-modification domain with BRCT motif and a WGR domain having conserved tryptophan, glycine, and arginine residues, and a C-terminal catalytic domain with PARP signature sequence.

Application

PARP1 Active human has been used in GST-PARP1 pulldown, to study the alterations in protein-protein interactions of sex determining region Y-box 2 (SOX2) upon O-GlcNAcylation. It has also been used in in vitro PARylation assay to investigate the effect of NR1D1 on DNA repair after damage induced by doxorubicin in breast cancer cells. It is useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Biochem/physiol Actions

PARPs comprise a set of enzymes that govern cellular processes such as DNA damage response, cell metabolism, chromatin remodeling, and transcriptional regulation. PARP1 detects and mends DNA breaks via base excision repair when DNA damage occurs. In the absence of PARP1 activity, damaged DNA accumulates, leading to impaired DNA replication.

Unit Definition

One unit of PARP incorporates 100 pmoles of poly(ADP) in 1 minute (room temperature) from NAD into acid-insoluble form.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 50% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proteolysis-targeting chimeras: A promising technique in cancer therapy for gaining insights into tumor development
Lv M, et al.
Cancer Letters, 539, 215716-215716 (2022)
Anneli Andersson et al.
Nucleic acids research, 44(16), 7630-7645 (2016-05-21)
Harmful oxidation of proteins, lipids and nucleic acids is observed when reactive oxygen species (ROS) are produced excessively and/or the antioxidant capacity is reduced, causing 'oxidative stress'. Nuclear poly-ADP-ribose (PAR) formation is thought to be induced in response to oxidative
Kostantin Kiianitsa et al.
DNA repair, 96, 102977-102977 (2020-10-12)
The nucleoside analog 5-aza-2'-deoxycytidine (5-aza-dC) is used to treat some hematopoietic malignancies. The mechanism of cell killing depends upon DNMT1, but is otherwise not clearly defined. Here we show that PARP1 forms covalent DNA adducts in human lymphoblast or fibroblasts
Fengxiao Zhang et al.
International journal of biological sciences, 16(15), 2868-2882 (2020-10-17)
Liver X receptor α (LXRα) controls a set of key genes involved in cholesterol metabolism. However, the molecular mechanism of this regulation remains unknown. The regulatory role of poly(ADP-ribose) polymerase 1 (PARP1) in cholesterol metabolism in the liver was examined.
Caifeng Liu et al.
Journal of integrative plant biology, 59(7), 459-474 (2017-03-07)
Root organogenesis involves cell division, differentiation and expansion. The molecular mechanisms regulating root development are not fully understood. In this study, we identified poly(adenosine diphosphate (ADP)-ribose) polymerases (PARPs) as new players in root development. PARP catalyzes poly(ADP-ribosyl)ation of proteins by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service