Skip to Content
Merck
All Photos(3)

Key Documents

SRE0045

Sigma-Aldrich

Luciferase from Photinus pyralis (firefly)

recombinant, expressed in E. coli, lyophilized powder, ≥10×1010 units/mg protein

Synonym(s):

Luciferase firefly

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

form

lyophilized powder

specific activity

≥10×1010 units/mg protein

mol wt

62 kDa

application(s)

diagnostic assay manufacturing

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Firefly luciferase is used extensively in molecular and cell biology, in particular for the efficient detection and quantitation of ATP and as a reporter for genetic function.

Biochem/physiol Actions

Firefly luciferase is a 62 kDa protein that catalyzes the production of light. The enzyme requires ATP, molecular oxygen, and the heterocyclic compound, firefly luciferin, to generate light in a two-step process. The light producing reaction is initiated by luciferin activation (adenylation of its carboxylate group) and proceeds in the presence of molecular oxygen to yield a photon of yellow-green light.

Unit Definition

One luciferase enzyme unit will produce one Relative Light Unit (RLU) at 20-25 °C over a 10 second period, measured in 100 μl assay mixture containing 40 pmol ATP and 15 nmol luciferin in Tris-glycine buffer, pH 7.6, using a GloMax 20/20 Luminometer.

Unit Definition Conversion Factor: There are approximately 9000 Relative Light Units (RLU) per one traditional Light Unit that uses a peak height equivalent to 0.02 μCi of 14C in a PPO/POPOP cocktail.

Physical form

Supplied as a lyophilized powder containing HEPES, pH 7.5, NaCl, MgCl2, EDTA, DTT and a carbohydrate stabilizer.
This product is a recombinant luciferase (62 kDa) from Photinus pyralis (American firefly) produced from the luc gene expressed in E. coli.

Preparation Note

To obtain maximal solubility it is important to reconstitute the enzyme at a high salt concentration, such as 1 M Tris buffer with any counter ion at pH 7-8. The enzyme can be prepared at a concentration of up to 5 mg protein/ml. Do not vortex and avoid agitation.
After reconstitution, the enzyme solutions can kept at 4-8 °C for up to 2 days or frozen in working aliquots at -20°C for at least one month. Repeated freezing and thawing is not recommended.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

S R Ford et al.
Journal of bioluminescence and chemiluminescence, 7(3), 185-193 (1992-07-01)
Commercially available crystalline native and recombinant firefly luciferases were compared. The two types of luciferase had indistinguishable responses to variation in ATP and luciferin concentrations and to omission of reaction components. The time courses of light production, the responses to
Pratik Vyas et al.
Journal of the American Chemical Society, 145(15), 8344-8354 (2023-03-24)
Reactions involving the transfer of a phosphoryl (-PO32-) group are fundamental to cellular metabolism. These reactions are catalyzed by enzymes, often large and complex, belonging to the phosphate-binding loop (P-loop) nucleoside triphosphatase (NTPase) superfamily. Due to their critical importance in
Kanika Narula et al.
The Plant journal : for cell and molecular biology, 103(2), 561-583 (2020-03-15)
Pathogen-/microbe-associated molecular patterns (PAMPs/MAMPs) initiate complex defense responses by reorganizing the biomolecular dynamics of the host cellular machinery. The extracellular matrix (ECM) acts as a physical scaffold that prevents recognition and entry of phytopathogens, while guard cells perceive and integrate
Maura Greiser et al.
eLife, 12 (2023-06-05)
Mitochondrial ATP production in ventricular cardiomyocytes must be continually adjusted to rapidly replenish the ATP consumed by the working heart. Two systems are known to be critical in this regulation: mitochondrial matrix Ca2+ ([Ca2+]m) and blood flow that is tuned
Aleksey Yevtodiyenko et al.
Nature communications, 12(1), 2680-2680 (2021-05-13)
Bioluminescent imaging (BLI) is one of the most powerful and widely used preclinical imaging modalities. However, the current technology relies on the use of transgenic luciferase-expressing cells and animals and therefore can only be applied to a limited number of

Articles

Firefly luciferase is a sensitive reporter for gene studies due to its absence in mammalian cells or tissues.

Firefly luciferase is a sensitive reporter for gene studies due to its absence in mammalian cells or tissues.

Firefly luciferase is a sensitive reporter for gene studies due to its absence in mammalian cells or tissues.

Firefly luciferase is a sensitive reporter for gene studies due to its absence in mammalian cells or tissues.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service