Skip to Content
Merck
All Photos(1)

Key Documents

SML2760

Sigma-Aldrich

SGC6870

≥98% (HPLC)

Synonym(s):

(R)-4-(5-Bromothiophene-2-carbonyl)-5-(3,5-dimethylphenyl)-7-methyl-4,5-dihydro-1H-benzo[e][1,4]diazepin-2(3H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H21BrN2O2S
Molecular Weight:
469.39
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +260 to +300°, c = 0.5 in methanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

SGC6870 is a potent and selective protein arginine methyltransferase PRMT6 inhibitor (IC50 = 77 nM by cell-free assays; IC50 = 0.8 μM against asymmetric H3R2 dimethylation of overexpressed PRMT6 in HEK293T cells) over all other PRMTs and 23 methyltransferases. The (S)-enantiomer SGC6870N is inactive toward PRMT6 and serves as an excellent control compound.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service