Skip to Content
Merck

SML2272

AZD-2098

≥98% (HPLC)

Synonym(s):

2,3-Dichloro-N-(3-methoxy-2-pyrazinyl)benzenesulfonamide, 2,3-Dichloro-N-(3-methoxypyrazin-2-yl)benzenesulfonamide, AZD 2098, AZD2098

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H9Cl2N3O3S
CAS Number:
Molecular Weight:
334.18
UNSPSC Code:
51111800
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=S(C1=CC=CC(Cl)=C1Cl)(NC2=NC=CN=C2OC)=O

InChI key

FLSMVCMSUNISFK-UHFFFAOYSA-N

Biochem/physiol Actions

AZD-2098 is an orally active, potent and selective CCR4 antagonist (pIC50 = 7.8/8.0/8.0/7.6 against CCL22 binding to human/mouse/rat/dog CCR4; inactive toward CXCR1/2 & CCR1/2b/5/7/8 at 10 μM) with little or no activity toward a panel of ∼120 receptors/enzymes. AZD-2098 effectively inhibits chemokine-induced cellular responses in vitro (pIC50 = 7.5 against CCL22-induced calcium influx in hCCR4-expressing CHO cells; pIC50 = 6.3 against CCL17- or CCL22-induced chemotaxis of primary human Th2 cells) and exhibits efficacy against antigen-induced inflammatory response among ovalbumin-sensitized rats in vivo (Emax = 7.5 μmol/kg p.o.; 1 h before and every 12 h after antigen challenge).
Orally active, potent and selective CCR4 antagonist with efficacy against antigen-induced inflammatory response in ovalbumin-sensitized rats in vivo.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library