Skip to Content
Merck

SML2223

WAY-267464 dihydrochloride

≥98% (HPLC)

Synonym(s):

4-(3,5-Dihydroxy-benzyl)-piperazine-1-carboxylic acid 2-methyl-4-(3-methyl-4,10-dihydro-3H-2,3,4,9-tetraaza-benzo[f]azulene-9-carbonyl)-benzylamide dihydrochloride, N-[[4-[(4,10-Dihydro-1-methylpyrazolo[3,4-b][1,5]benzodiazepin-5(1H)-yl)carbonyl]-2-methylphenyl]methyl]-4-[(3,5-dihydroxyphenyl)methyl]-1-piperazinecarboxamide dihydrochloride, WAY 267,464 dihydrochloride, WAY 267464 dihydrochloride, WAY-267,464 dihydrochloride, WAY267464 dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C32H35N7O4 · 2HCl
CAS Number:
Molecular Weight:
654.59
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NCC1=CC=C(C=C1C)C(N2CC3=C(NC4=CC=CC=C24)N(N=C3)C)=O)N5CCN(CC5)CC6=CC(O)=CC(O)=C6

InChI

1S/C32H35N7O4.2ClH/c1-21-13-23(31(42)39-20-25-18-34-36(2)30(25)35-28-5-3-4-6-29(28)39)7-8-24(21)17-33-32(43)38-11-9-37(10-12-38)19-22-14-26(40)16-27(41)15-22;;/h3-8,13-16,18,35,40-41H,9-12,17,19-20H2,1-2H3,(H,33,43);2*1H

InChI key

OTFWXMFLPMUDFP-UHFFFAOYSA-N

Biochem/physiol Actions

Non-peptide oxytocin receptor (OTR) agonist and vasopressin V1a receptor (V1aR) antagonist.
WAY-267464 is a non-peptide drug with a novel mechanism of action (MOA) to treat psychosis and schizophrenia.
WAY-267464 is a non-peptide oxytocin receptor (OTR) agonist (EC50 = 61-881 nM; Ki = 58-978 nM) that, unlike oxytocin (OT), displays antagonist instead of agonist activity toward vasopressin V1a receptor/V1aR (IC50 = 613 nM; Ki = 27-113 nM). WAY-267464 exhibits OT-like anxiolytic effects in assays measuring both behavioral (33% increase in punished crossing by 10 mg/mL ip or 10 μg/mouse icv in four-plate tests; 75% increased open quadrants stay by 3 μg/mouse icv in elevated zero maze) and autonomic (47% higher stress-induced hyperthermia by 10 μg/mouse icv) parameters of the anxiety response. Similar to the antipsychotic-like effects reported for OT, WAY-267464 also reverses disruption in prepulse inhibition of the acoustic startle reflex induced by either MK-801 or amphetamine. Unlike OT, WAY-267464 does not affect immobility in mouse tail suspension test.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library