Skip to Content
Merck
All Photos(1)

Documents

SML1333

Sigma-Aldrich

Pimobendan

≥98% (HPLC)

Synonym(s):

(RS)-6-[2-(4-Methoxyphenyl)-1H-benzimidazol-5-yl]-5-methyl-4,5-dihydropyridazin-3(2H)-one, 4,5-Dihydro-6-[2-(4-methoxyphenyl)-1H-benzimidazol-5-yl]-5-methyl-3(2H)-pyridazinone, 4,5-Dihydro-6-[2-(4-methoxyphenyl)-1H-benzimidazol-6-yl]-5-methyl-3(2H)-pyridazinone, 4,5-Dihydro-6-[2-(4-methoxyphenyl)-1H-benzimidazol-6-yl]-5-methyl-3(2H)-pyridazinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18N4O2
CAS Number:
Molecular Weight:
334.37
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CC(C(C1=CC=C(NC(C2=CC=C(OC)C=C2)=N3)C3=C1)=NN4)CC4=O

InChI

1S/C19H18N4O2/c1-11-9-17(24)22-23-18(11)13-5-8-15-16(10-13)21-19(20-15)12-3-6-14(25-2)7-4-12/h3-8,10-11H,9H2,1-2H3,(H,20,21)(H,22,24)

InChI key

GLBJJMFZWDBELO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Pimobendan is an inotropic agent with a dual mechanism of action: it increases myocardial contractility by increasing calcium sensitization to troponin C and it promotes vasodilation by inhibiting phosphodiesterase III (PDE3). Pimobendan has been studied for treating heart failure and cardiomyopathy, primarily for veterinary uses.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stage-dependent benefits and risks of pimobendan in mice with genetic dilated cardiomyopathy and progressive heart failure.
Nonaka M, Morimoto S, Murayama T, et al.
British Journal of Pharmacology, 172(9), 2369-2382 (2015)
Telma M Nakata et al.
Journal of cardiovascular pharmacology, 65(6), 640-648 (2015-03-26)
This study was designed to assess the progression of pulmonary arterial hypertension (PAH) and the effectiveness of therapy using recently investigated echocardiographic parameters. PAH is characterized by the progressive elevation of pulmonary artery pressure and right ventricular hypertrophy and dysfunction
Kimberly L Boyle et al.
Journal of veterinary emergency and critical care (San Antonio, Tex. : 2001), 22(4), 398-408 (2012-08-30)
To review the pharmacology, research developments, and clinical uses of pimobendan Original research articles and clinical studies from 1984 to August 2011. Pimobendan is approved for use in dogs for the treatment of congestive heart failure (CHF) secondary to chronic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service