Skip to Content
Merck
All Photos(1)

Key Documents

SML0804

Sigma-Aldrich

(S)-CPP sodium salt

≥96% (HPLC)

Synonym(s):

(S)-2-Chloro-3-phenylpropanoic acid sodium salt, (aS)-a-chloro-benzenepropanoic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8ClO2 · Na
CAS Number:
Molecular Weight:
206.60
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥96% (HPLC)

form

powder

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

[O-]C([C@@H](Cl)CC1=CC=CC=C1)=O.[Na+]

InChI

1S/C9H9ClO2.Na/c10-8(9(11)12)6-7-4-2-1-3-5-7;/h1-5,8H,6H2,(H,11,12);/q;+1/p-1/t8-;/m0./s1

InChI key

HMHPCHNNVOLTFC-QRPNPIFTSA-M

Biochem/physiol Actions

(S)-CPP is an inhibitor of branched-chain α-ketoacid dehydrogenase complex (BCKDC) kinase, also know as BDK, or keto acid dehydrogenase kinase. BDK is a negative regulator of BCKDC activity. Inhibition of BDK by (S)-CPP (IC50 = 6.3 μM) leads to complex activation, and significant reduction of plasma levels of leucine/isoleucine and valine in wild type mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service